Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLGAP2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-294 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-294 Supplier Abnova Corporation Supplier No. PAB13041
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DLGAP2.

The product of this gene is one of the membrane-associated guanylate kinases localized at postsynaptic density in neuronal cells. These kinases are a family of signaling molecules expressed at various submembrane domains and contain the PDZ, SH3 and the guanylate kinase domains. This protein may play a role in the molecular organization of synapses and in neuronal cell signaling. Alternatively spliced transcript variants encoding different isoforms have been identified, but their full-length nature is not known. [provided by RefSeq

Specifications

Antigen DLGAP2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DLGAP2.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene DLGAP2
Gene Accession No. Q9P1A6
Gene Alias DAP2/SAPAP2
Gene Symbols DLGAP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 16 amino acids of human DLGAP2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9228
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.