Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMKN, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123988 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-988 Supplier Abnova Corporation Supplier No. PAB22282
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DMKN.

This gene is upregulated in inflammatory diseases, and it was first observed as expressed in the differentiated layers of skin. The most interesting aspect of this gene is the differential use of promoters and terminators to generate isoforms with unique cellular distributions and domain compliments. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene, but the full-length nature of some of them has not been determined. [provided by RefSeq

Sequence: GGNGHKPGNSETSPGMFNFDTFWKNFKSKLGFINWDAINKNQVPPPSTRALLYFSRLWEDFKQNTPFLNWKAIIEGADASSLQKRAGRADQNYNYNQHAYPTAYGGKYSVKTPAKGGVSPSSSASR

Specifications

Antigen DMKN
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DMKN.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DMKN
Gene Accession No. Q6E0U4
Gene Alias UNQ729/ZD52F10
Gene Symbols DMKN
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DMKN.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93099
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.