Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-DMRT2, Polyclonal,

Catalog No. ABE1364MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1364MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1364MI Supplier MilliporeSigma™ Supplier No. ABE1364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects DMRT2 in Human, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Doublesex- and mab-3-related transcription factor 2 is encoded by the DMRT2 gene in human and directly regulates early activation of the myogenic determination gene MYF5 by binding in a sequence-specific manner to its early epaxial enhancer element. In addition, it is also involved in somite develoment during embryogenesis. The human protein shares a DM DNA-binding domain with Drosophila 'doublesex' (dsx) and C. elegans mab3 gene products involved in sex determination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9Y5R5
Gene Symbols DMRT2; DSXL2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human DMRT2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_870987
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.