Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-DNA polymerase beta, Polyclonal,

Catalog No. ABE1408MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1408MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1408MI Supplier MilliporeSigma™ Supplier No. ABE1408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects DNA polymerase beta in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

DNA polymerase beta (EC 2.7.7.7; UniProt P06746; also known as DNA pol beta, DNA polymerase beta subunit) is encoded by the POLB gene (Gene ID 5423) in human. DNA polymerase beta is involved in DNA recombination and base-excision repair (BER). Upon oxidative DNA damage, Polb translocates to the nucleus, where it associates with AP endonuclease 1 (Apex) to carry out DNA repair. Apex makes a single nick 5′ to an abasic site in double-stranded DNA, Polb then inserts the replacement nucleotide(s). Aberrant POLB overexpression results in an increased rate of spontaneous mutagenesis and a highly mutagenic tolerance phenotype. Thirty percent of all tumors are reported to harbor mutations in the POLB gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA polymerase beta
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P06746
Gene Symbols POLB
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human DNA polymerase beta.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002681
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.