Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAH17, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-997 Supplier Abnova Corporation Supplier No. PAB27485

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against recombinant DNAH17.

Dyneins are microtubule-associated motor protein complexes composed of several heavy, light, and intermediate chains. DNAH17 is a heavy chain associated with axonemal dynein (Milisav and Affara, 1998 [PubMed 9545504]).[supplied by OMIM

Sequence: VFQYIQEVREILHNLQNRMQKAKQNIEGISQAMKDWSANPLFERKDNKKEALLDLDGRIANLNKRYAAVRDAGVKIQAMVAVRKHPG

Specifications

Antigen DNAH17
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description dynein, axonemal, heavy chain 17
Formulation In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
Gene DNAH17
Gene Alias DNAHL1, DNEL2, FLJ40457, MGC132767, MGC138489
Gene Symbols DNAH17
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DNAH17.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8632
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.