Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAH8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123861 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-861 Supplier Abnova Corporation Supplier No. PAB22154
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DNAH8.

Dyneins are microtubule-associated motor protein complexes composed of several heavy, light, and intermediate chains. Dynein heavy chains (DHCs) are responsible for force production and ATPase activity and contain a highly conserved catalytic domain with 4 P-loop consensus motifs involved in nucleotide binding. Two major classes of dyneins, axonemal and cytoplasmic, have been identified. Axonemal dyneins, found in cilia and flagella, are components of the outer and inner dynein arms attached to the peripheral microtubule doublets. DNAH8 is an outer arm axonemal DHC (Chapelin et al., 1997 [PubMed 9256245]; Neesen et al., 1997 [PubMed 9373155]).[supplied by OMIM

Sequence: ILNHKSKHVEEAVRELISIFEQIYEVKYTGKVGKQSEQRKHVVFGSETEEGENNDYEANIVNEFDTHDKEDEFKKECKEVFAFFSHQLLDSLQKATRLSLD

Specifications

Antigen DNAH8
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DNAH8.
Dilution Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DNAH8
Gene Accession No. Q96JB1
Gene Alias ATPase/FLJ25850/FLJ36115/FLJ36334/hdhc9
Gene Symbols DNAH8
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DNAH8.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1769
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.