Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-603 Supplier Abnova Corporation Supplier No. PAB15411
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DNAJB1.

Specifications

Antigen DNAJB1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DNAJB1.
Dilution Western Blot (1:1500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene DNAJB1
Gene Alias HSPF1/Hdj1/Hsp40/Sis1
Gene Symbols DNAJB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human DNAJB1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3337
Test Specificity Detects an ∼40KDa protein corresponding to the molecular mass of Hsp40 on SDS PAGE immunoblots. Also detects recombinant Hsp40.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.