Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt1 Rabbit anti-Frog, Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117665 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-665 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-665 Supplier Abnova Corporation Supplier No. PAB15591
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Dnmt1.

Specifications

Antigen Dnmt1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Dnmt1.
Dilution Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene Dnmt1
Gene Alias Cxxc9/Dnmt/Dnmt1o/MTase/Met-1/Met1/MommeD2
Gene Symbols Dnmt1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1037-1386 mouse Dnmt1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13433
Target Species Frog, Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.