Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt3l, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124006 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-006 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-006 Supplier Abnova Corporation Supplier No. PAB2230
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Dnmt3l.

Specifications

Antigen Dnmt3l
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Dnmt3l.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Dnmt3l
Gene Accession No. Q9CWR8
Gene Alias D6Ertd14e/MGC102308/ecat7
Gene Symbols Dnmt3l
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to the last 100 amino acids of mouse Dnmt3l.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54427
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.