Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dock4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117791 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-791 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-791 Supplier Abnova Corporation Supplier No. PAB15726
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Dock4.

Sequence: CLSPRDRPCSAIYPTPVEPSQRMLFNHIGDGALPRSDPNLSAPEKAVNPTPSSWSLDSGKEAKNMSDSGKLISPPVPPRPTQTASPARHTTSVSPSPAGRSPLKGSVQSFTPSPVEYNSPGLSSNSPVLSGSYSSGISSLSRCSTSETSGFENQANEQSVPVPVPVPVPVPVPSFSGSEEPVRKESKTPPPYSVYERTLRRPVPLPHSLSIPVTSEPPALPPKPLAARSSHLENGTRRTEPGPRPRPLPRKVSQ

Specifications

Antigen Dock4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Dock4.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Dock4
Gene Accession No. AK122353
Gene Alias 5330406C03/6330411N01Rik/AF263288/C030023J22/mKIAA0716
Gene Symbols Dock4
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 254 mouse Dock4.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238130
Test Specificity Specific to recombinant protein GX0390. This antibody detects mDOCK4 protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.