Learn More
DOK1 (phospho S450), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of DOK1.
Supplier: Abnova Corporation PAB7908
Description
Docking protein 1 is constitutively tyrosine phosphorylated in hematopoietic progenitors isolated from chronic myelogenous leukemia (CML) patients in the chronic phase. It may be a critical substrate for p210(bcr/abl), a chimeric protein whose presence is associated with CML. Docking protein 1 contains a putative pleckstrin homology domain at the amino terminus and ten PXXP SH3 recognition motifs. Docking protein 2 binds p120 (RasGAP) from CML cells. It has been postulated to play a role in mitogenic signaling. [provided by RefSeq
Specifications
| DOK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of DOK1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| MGC117395/MGC138860/P62DOK | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This sequence is conserved in Dok1 from rat (Ser-449) and mouse (Ser-451). The site is not conserved in other Dok family members. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| DOK1 | |
| DOK1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S450 of human DOK1. | |
| RUO | |
| 1796 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only