Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPEP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89127703 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-703 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-703 Supplier Abnova Corporation Supplier No. PAB26064
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DPEP1.

DPEP1 (EC 3.4.13.11) is a kidney membrane enzyme that hydrolyzes a variety of dipeptides and is implicated in renal metabolism of glutathione and its conjugates, e.g., leukotriene D4 (Kozak and Tate, 1982 [PubMed 6122685]). DPEP1 is responsible for hydrolysis of the beta-lactam ring of antibiotics, such as penem and carbapenem (Campbell et al., 1984 [PubMed 6334084]). Earlier, beta-lactamase enzymes were thought to occur only in bacteria, where their probable function was in protecting the organisms against the action of beta-lactam antibiotics. These antibiotics exhibit selective toxicity against bacteria but virtual inertness against many eukaryotic cells (Adachi et al., 1990 [PubMed 2303490]).[supplied by OMIM

Specifications

Antigen DPEP1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DPEP1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene DPEP1
Gene Accession No. P16444
Gene Alias MBD1/MDP/RDP
Gene Symbols DPEP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at N-terminus of human DPEP1.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1800
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.