Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPH3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-383 Supplier Abnova Corporation Supplier No. PAB22674
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DPH3.

This gene encodes a CSL zinc finger-containing protein that is required for dipthamide biosynthesis. The encoded protein is necessary for the initial step in the modification of a histidine residue in elongation factor-2 to diphthamide. This modified residue is a target for ADP ribosylation by the bacterial toxins diphtheria toxin and Pseudomonas exotoxin A. Alternative splicing results in multiple transcript variants that encode the same isoform. [provided by RefSeq

Sequence: EDFQYDEDSETYFYPCPCGDNFSITKEDLENGEDVATCPSCSLIIKVIYDKDQFVCGETVPAPSANKELVK

Specifications

Antigen DPH3
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DPH3.
Dilution Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DPH3
Gene Accession No. Q96FX2
Gene Alias DELGIP/DELGIP1/DESR1/DPH3A/KTI11/MGC20197/ZCSL2
Gene Symbols DPH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DPH3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285381
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.