Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYSL2 (phospho T509), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89130674 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-674 Supplier Abnova Corporation Supplier No. PAB29239
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human DPYSL2.

Specifications

Antigen DPYSL2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human DPYSL2.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene DPYSL2
Gene Accession No. Q16555
Gene Alias CRMP2/DHPRP2/DRP-2/DRP2
Gene Symbols DPYSL2
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T509 of human DPYSL2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1808
Test Specificity This antibody detects endogenous levels of DPYSL2 only when phosphorylated at threonine 509.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.