Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRG2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-892 Supplier Abnova Corporation Supplier No. PAB20391
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DRG2.

The DRG2 gene encodes the developmentally regulated GTP-binding protein 2, a name derived from the fact that it shares significant similarity to known GTP-binding proteins. DRG2 was identified because it is expressed in normal fibroblasts but not in SV40-transformed fibroblasts. Read-through transcripts containing this gene and a downstream gene have been identified, but they are not thought to encode a fusion protein. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq

Sequence: YKIFNAEVLFREDCSPDEFIDVIVGNRVYMPCLYVYNKIDQISMEEVDRLARKPNSVVISCGMKLNLDYLLEMLWEYLALTCIYTKKRGQRPDFTDAIILRKGASVEHVCHRIHRSLASQFKYALVWGTSTKYSPQRVGLTHTMEHEDVI

Specifications

Antigen DRG2
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DRG2.
Dilution Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DRG2
Gene Accession No. P55039
Gene Symbols DRG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DRG2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1819
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.