Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRP2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121645 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-645 Supplier Abnova Corporation Supplier No. PAB20143
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant DRP2.

Members of the dystrophin family of proteins perform a critical role in the maintenance of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord. [provided by RefSeq

Sequence: SRIEHFASRLAEMESQNCSFFNDSLSPDDSIDEDQYLLRHSSPITDREPAFGQQAPCSVATESKGELQKILAHLEDENRILQGELRRLKWQHEEAAEAPSLADGSTEAATDHRNEELLAEARILRQHKSRLETRMQILEDHNKQLESQ

Specifications

Antigen DRP2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DRP2.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DRP2
Gene Accession No. Q13474
Gene Alias MGC133255
Gene Symbols DRP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DRP2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1821
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.