Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSG4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89131818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-818 Supplier Abnova Corporation Supplier No. PAB5325
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DSG4.

This gene encodes a member of the desmoglein subgroup of desmosomal cadherins. The encoded protein is a transmembrane component in desmosomes and may play a role in cell-cell adhesion in epithelial cells. Mutations in the gene are associated with localized autosomal recessive hypotrichosis and potentially in other skin disorders. Alternate splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen DSG4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DSG4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene DSG4
Gene Alias CDGF13/CDHF13/LAH
Gene Symbols DSG4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human DSG4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147409
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.