Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP19 Rabbit, Polyclonal Antibody, Abnova™
Supplier: Abnova Corporation PAB14631
Specifications
DUSP19 | |
Polyclonal | |
In serum (50% glycerol, 0.025% sodium azide) | |
DUSP19 | |
A synthetic peptide corresponding to amino acids 74-89 of human DUSP19. | |
Primary | |
Bovine, Chicken, Human, Mouse | |
Serum |
ELISA | |
Unconjugated | |
NP_543152;Q8WTR2 | |
Rabbit | |
100 μL | |
142679 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction