Learn More
DUSP4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DUSP4.
Supplier: Abnova Corporation PAB4129
Description
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus. Two alternatively spliced transcript variants, encoding distinct isoforms, have been observed for this gene. In addition, multiple polyadenylation sites have been reported. [provided by RefSeq
Specifications
DUSP4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of DUSP4. | |
In PBS (0.09% sodium azide) | |
NP_001385;Q13115 | |
DUSP4 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DUSP4. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
DUSP4 | |
HVH2/MKP-2/MKP2/TYP | |
Rabbit | |
Protein G purification | |
RUO | |
1846 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only