Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYNLRB2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-366 Supplier Abnova Corporation Supplier No. PAB17702
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DYNLRB2.

Specifications

Antigen DYNLRB2
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DYNLRB2.
Dilution Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene DYNLRB2
Gene Accession No. Q8TF09
Gene Alias DNCL2B/DNLC2B/MGC62033/ROBLD2
Gene Symbols DYNLRB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human DYNLRB2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83657
Test Specificity This antibody detects endogenous levels of total DYNLRB2 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.