Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2f3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117865 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-865 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-865 Supplier Abnova Corporation Supplier No. PAB15802
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant E2f3.

Specifications

Antigen E2f3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant E2f3.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene E2f3
Gene Accession No. AK129048
Gene Alias E2F3b/E2f3a/mKIAA0075
Gene Symbols E2f3
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 165 amino acids of mouse E2f3.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13557
Test Specificity Specific to recombinant protein GX1865. This antibody detects mE2F3 protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.