Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F4/E2F5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128705 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-705 Supplier Abnova Corporation Supplier No. PAB27055
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of E2F4/E2F5.

Specifications

Antigen E2F4/E2F5
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of E2F4/E2F5.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene Accession No. Q16254 (Gene ID : 1874);Q15329 (Gene : 1875)
Gene Alias E2F-4
Gene Symbols E2F4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human E2F4/E2F5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1874/1875
Test Specificity E2F4/E2F5 polyclonal antibody detects endogenous levels of E2F4/E2F5 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.