Learn More
ECT2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ECT2.
Supplier: Abnova Corporation PAB10006
Description
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis. [provided by RefSeq
Specifications
ECT2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ECT2. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
NP_060568;Q9H8V3 | |
ECT2 | |
Synthetic phosphopeptide corresponding to residues surrounding T790 of human ECT2. | |
RUO | |
1894 | |
Canine, Chicken, Chimpanzee, Fish, Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:10000-1:40000) Western Blot (1:1000-1:4000) The optimal working dilution should be determined by the end user. | |
ECT2 | |
FLJ10461/MGC138291 | |
Rabbit | |
100 μg | |
Primary | |
Reactivity occurs against human ECT2 protein at the pT790 residue. By ELISA This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated human ECT2 is minimal by ELISA, but western blot shows specific staining of interphase cell lysates, which are predicted to contain non-phosphorylated ECT2. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only