Learn More
EDNRB Rabbit anti-Dog, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EDNRB.
Supplier: Abnova Corporation PAB26263
Description
The protein encoded by this gene is a G protein-coupled receptor which activates a phosphatidylinositol-calcium second messenger system. Its ligand, endothelin, consists of a family of three potent vasoactive peptides: ET1, ET2, and ET3. Studies suggest that the multigenic disorder, Hirschsprung disease type 2, is due to mutations in the endothelin receptor type B gene. Three transcript variants encoding two different isoforms have been found for this gene. While both isoforms bind ET1, they exhibit different responses upon binding, suggesting that they may be functionally distinct. [provided by RefSeq
Specifications
EDNRB | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of EDNRB. | |
In PBS (0.09% sodium azide) | |
P24530 | |
EDNRB | |
A synthetic peptide corresponding to 18 amino acid at N-terminus of human EDNRB. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16 ug/mL) The optimal working dilution should be determined by the end user. | |
EDNRB | |
ABCDS/ETB/ETBR/ETRB/HSCR/HSCR2 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
1910 | |
Canine, Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only