Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDNRB Rabbit anti-Dog, Human, Polyclonal Antibody, Abnova™

Catalog No. 89127860 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-860 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-860 Supplier Abnova Corporation Supplier No. PAB26220
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EDNRB.

The protein encoded by this gene is a G protein-coupled receptor which activates a phosphatidylinositol-calcium second messenger system. Its ligand, endothelin, consists of a family of three potent vasoactive peptides: ET1, ET2, and ET3. Studies suggest that the multigenic disorder, Hirschsprung disease type 2, is due to mutations in the endothelin receptor type B gene. Three transcript variants encoding two different isoforms have been found for this gene. While both isoforms bind ET1, they exhibit different responses upon binding, suggesting that they may be functionally distinct. [provided by RefSeq

Specifications

Antigen EDNRB
Applications Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EDNRB.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-11 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene EDNRB
Gene Accession No. P24530
Gene Alias ABCDS/ETB/ETBR/ETRB/HSCR/HSCR2
Gene Symbols EDNRB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 19 amino acid at N-terminus of human EDNRB.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1910
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except PTPN5 (68%).
Target Species Canine, Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.