Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF1E1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130026 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-026 Supplier Abnova Corporation Supplier No. PAB28516
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant EEF1E1.

This gene encodes a multifunctional protein that localizes both in the cytoplasm and in the nucleus. In the cytoplasm, the encoded protein is an auxiliary component of the macromolecular aminoacyl-tRNA synthase complex. However, its mouse homolog has been shown to translocate to the nucleus in response to DNA damage and plays a positive role in ATM/ATR-mediated p53 activation. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen EEF1E1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant EEF1E1.
Dilution Western Blot (1:8000) Immunohistochemistry Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene EEF1E1
Gene Alias AIMP3/P18
Gene Symbols EEF1E1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human EEF1E1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9521
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.