Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFEMP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-595 Supplier Abnova Corporation Supplier No. PAB16759
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EFEMP1.

This gene spans approximately 18kb of genomic DNA and consists of 12 exons. Alternative splice patterns in the 5' UTR result in three transcript variants encoding the same extracellular matrix protein. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. [provided by RefSeq]

Specifications

Antigen EFEMP1
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EFEMP1.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene EFEMP1
Gene Accession No. NP_004096
Gene Alias DHRD/DRAD/FBLN3/FBNL/FLJ35535/MGC111353/MLVT/MTLV/S1-5
Gene Symbols EFEMP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 19 amino acids of human EFEMP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2202
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.