Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFNB1/EFNB2/EFNB3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-667 Supplier Abnova Corporation Supplier No. PAB29232
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EFNB1/EFNB2/EFNB3.

Specifications

Antigen EFNB1/EFNB2/EFNB3
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EFNB1/EFNB2/EFNB3.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene Accession No. P98172;P52799;Q15768
Gene Alias CFND/CFNS/EFL3/EPLG2/Elk-L/LERK2/MGC8782
Gene Symbols EFNB1
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y324 of human EFNB1/EFNB2/EFNB3.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1947/1948/1949
Test Specificity This antibody detects endogenous levels of EFNB1/EFNB2/EFNB3 only when phosphorylated at tyrosine 324.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.