Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR, Rabbit, Clone: SP9, Abnova™

Catalog No. 89110825 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-825 Supplier Abnova Corporation Supplier No. MAB2118

Rabbit monoclonal antibody raised against synthetic peptide of EGFR.

The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq

Specifications

Antigen EGFR
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP9
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of EGFR.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Flow Cytometry (1:100) The optimal working dilution should be determined by the end user.
Formulation Tissue culture supernatant in TBS buffer, pH 7.5 (1% BSA, 0.09% sodium azide).
Gene EGFR
Gene Alias ERBB/ERBB1/HER1/PIG61/mENA
Gene Symbols EGFR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human EGFR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.