Learn More
EGFR (phospho S967), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.
Supplier: Abnova Corporation PAB7911
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Specifications
EGFR | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
ERBB/ERBB1/HER1/PIG61/mENA | |
Rabbit | |
100 μL | |
Primary | |
This human EGFR sequence has high homology to rat and mouse EGFR, and low homology to other ErbB family members. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
ELISA (1:2000) Immunocytochemistry (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
EGFR | |
EGFR | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S967 of human EGFR. | |
RUO | |
1956 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only