Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117140 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-140 Supplier Abnova Corporation Supplier No. PAB14556
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EGLN3.

Specifications

Antigen EGLN3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EGLN3.
Dilution Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7-8 (0.09% sodium azide)
Gene EGLN3
Gene Accession No. NP_071356.1
Gene Alias FLJ21620/HIFPH3/MGC125998/MGC125999/PHD3
Gene Symbols EGLN3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 50-100 of human EGLN3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112399
Test Specificity This antibody is specific to HIF prolyl hydroxylase 3.
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.