Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115813 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-813 Supplier Abnova Corporation Supplier No. PAB12439
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EGR2.

The protein encoded by this gene is a transcription factor with three tandem C2H2-type zinc fingers. Defects in this gene are associated with Charcot-Marie-Tooth disease type 1D (CMT1D), Charcot-Marie-Tooth disease type 4E (CMT4E), and with Dejerine-Sottas syndrome (DSS). Multiple transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen EGR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EGR2.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.09% sodium azide)
Gene EGR2
Gene Accession No. P11161
Gene Alias AT591/CMT1D/CMT4E/DKFZp686J1957/FLJ14547/KROX20
Gene Symbols EGR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of human EGR2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1959
Test Specificity This antibody is specific to EGR2.
Target Species Human, Pig
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.