Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2AK2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128445 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-445 Supplier Abnova Corporation Supplier No. PAB26802
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EIF2AK2.

Sequence: K-G-Tp-L-R

Specifications

Antigen EIF2AK2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EIF2AK2.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene EIF2AK2
Gene Accession No. P19525
Gene Alias EIF2AK1/MGC126524/PKR/PRKR
Gene Symbols EIF2AK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding T451 of human EIF2AK2.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5610
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.