Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eif2ak3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114680 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-680 Supplier Abnova Corporation Supplier No. PAB10175
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Eif2ak3.

Specifications

Antigen Eif2ak3
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Eif2ak3.
Dilution Western Blot (1:500-1:3000) Immunoprecipitation (10-30ul) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Eif2ak3
Gene Accession No. AAH54809;Q9Z2B5
Gene Alias AI427929/Pek/Perk
Gene Symbols Eif2ak3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 601-1115 of mouse Eif2ak3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13666
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.