Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eif3e, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-690 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-690 Supplier Abnova Corporation Supplier No. PAB10203
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Eif3e.

Specifications

Antigen Eif3e
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Eif3e.
Dilution ELISA (1:90000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Eif3e
Gene Accession No. NP_032414;P60229
Gene Alias 48kDa/Eif3s6/Int6/eIF3-p46/eIF3-p48
Gene Symbols Eif3e
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of mouse Eif3e.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16341
Test Specificity This antibody is specific to mouse eIF3S6/Int6 protein.
Target Species Bovine, Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.