Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF5A2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-620 Supplier Abnova Corporation Supplier No. PAB1009

Rabbit polyclonal antibody raised against synthetic peptide of EIF5A2.

Sequence: VREDLKLPEGELGKE

Specifications

Antigen EIF5A2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description eukaryotic translation initiation factor 5A2
Formulation In PBS (0.02% sodium azide)
Gene EIF5A2
Gene Alias EIF-5A2, eIF5AII
Gene Symbols EIF5A2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 108-122 of human EIF5A2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56648
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.