Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELK3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89129964 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-964 Supplier Abnova Corporation Supplier No. PAB28457
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ELK3.

The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present. [provided by RefSeq

Sequence: KNIIKKVIGQKFVYKFVSFPEILKMDPHAVEISRESLLLQDSDCKASPEGREAHKHGLAALRSTSRNEYIHSGLYSSFTINSLQNPPDAFKAIKTEKLEEPPEDSPPVEEVRTVIRFVTNKTDKHVTR

Specifications

Antigen ELK3
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ELK3.
Dilution Immunohistochemistry(1:500-1:1000) Western Blot(1:100-1:250) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene ELK3
Gene Accession No. P41970
Gene Alias ERP/NET/SAP2
Gene Symbols ELK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of recombinant ELK3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2004
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.