Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ELOVL1.
Supplier: Abnova Corporation PAB26984
Specifications
ELOVL1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
ELOVL1 | |
CGI-88/Ssc1 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
64834 | |
Human, Mouse | |
Liquid |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ELOVL1. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Q9BW60 | |
ELOVL1 | |
A synthetic peptide corresponding to human ELOVL1. | |
100 μL | |
Primary | |
ELOVL1 polyclonal antibody detects endogenous levels of ELOVL1 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction