Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL5 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89129347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-347 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-347 Supplier Abnova Corporation Supplier No. PAB27825
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ELOVL5.

ELOVL5 plays a role in elongation of long-chain polyunsaturated fatty acids (Leonard et al., 2000 [PubMed 10970790]).[supplied by OMIM

Specifications

Antigen ELOVL5
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ELOVL5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene ELOVL5
Gene Accession No. Q9NYP7
Gene Alias HELO1/dJ483K16.1
Gene Symbols ELOVL5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acid at C-terminus of human ELOVL5.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60481
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.