Learn More
EMCN Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant EMCN.
Supplier: Abnova Corporation PAB20327
Description
EMCN is a mucin-like sialoglycoprotein that interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix (Kinoshita et al., 2001 [PubMed 11418125]).[supplied by OMIM
Sequence: EAANNSLVVTTTKPSITTPNTESLQKNVVTPTTGTTPKGTITNELLKMSLMSTATFLTSKDEGLKATTTDVRKNDSIISNVTVTSVTLPNAVSTLQSSKPKTETQSSIKTTEIPSpecifications
| EMCN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant EMCN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9ULC0 | |
| EMCN | |
| Recombinant protein corresponding to amino acids of human EMCN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| EMCN | |
| EMCN2/MUC14 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51705 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only