Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMR2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89127832 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-832 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-832 Supplier Abnova Corporation Supplier No. PAB26192
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EMR2.

This gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 3 on chromosome 19. This protein does not interact with the ligand decay accelerating factor for complement, unlike the related CD97 antigen, and indicates that these very closely related proteins likely have nonredundant functions. This gene results in multiple transcripts encoding distinct isoforms. [provided by RefSeq

Specifications

Antigen EMR2
Applications ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EMR2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7.5-15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene EMR2
Gene Accession No. Q9UHX3
Gene Alias CD312/DKFZp781B135
Gene Symbols EMR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 20 amino acid at N-terminus of human EMR2.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30817
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except MYO18B (45%), FLJ13231 (40%).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.