Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPAS1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-622 Supplier Abnova Corporation Supplier No. PAB12124
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EPAS1.

Specifications

Antigen EPAS1
Applications ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EPAS1.
Dilution ChIP (1:10-1:500) ELISA (1:100-1:2000) Flow Cytometry Gel Supershift Assay Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sectio
Formulation In PBS (0.05% sodium azide)
Gene Accession No. P97481
Gene Alias ECYT4/HIF2A/HLF/MOP2/PASD2/bHLHe73
Gene Symbols EPAS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human and mouse EPAS1/Epas1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2034/13819
Test Specificity Specific for HIF-2 alpha/EPAS. Does not cross-react with HIF-1 alpha.
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.