Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epb41l1 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-814 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-814 Supplier Abnova Corporation Supplier No. PAB15749
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Epb4.1l1.

Specifications

Antigen Epb41l1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Epb4.1l1.
Dilution Western Blot (1:1000) Immunohistochemistry (1:25) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.05% sodium azide)
Gene Epb4.1l1
Gene Accession No. AB002336 (Human)
Gene Alias 4.1N
Gene Symbols Epb4.1l1
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 260 amino acids (L518-E777) of rat Epb4.1l1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59317
Test Specificity This antibody detects rat EPB41L1 protein.
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 200μL distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.