Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB49 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128776 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-776 Supplier Abnova Corporation Supplier No. PAB27123
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of EPB49.

Dematin, or EPB49, is an actin-bundling protein originally identified in the erythroid membrane skeleton. Its actin-bundling activity is abolished upon phosphorylation by cAMP-dependent protein kinase and is restored after dephosphorylation (Rana et al., 1993 [PubMed 8341682]).[supplied by OMIM

Specifications

Antigen EPB49
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of EPB49.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene EPB49
Gene Accession No. Q08495
Gene Alias DMT/FLJ78462/FLJ98848
Gene Symbols EPB49
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human EPB49.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2039
Test Specificity EPB49 polyclonal antibody detects endogenous levels of EPB49 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.