Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHA3/EPHA4/EPHA5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-735 Supplier Abnova Corporation Supplier No. PAB29297
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EPHA3/EPHA4/EPHA5.

Specifications

Antigen EPHA3/EPHA4/EPHA5
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EPHA3/EPHA4/EPHA5.
Dilution Immunohistochemistry (1:50∼1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Accession No. P29320;P54764;P54756
Gene Alias ETK/ETK1/HEK/HEK4/TYRO4
Gene Symbols EPHA3
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y779/Y833 of human EPHA3/EPHA4/EPHA5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2042/2043/2044
Test Specificity EPHA3/EPHA4/EPHA5 (phospho Y779/Y833) polyclonal antibody detects endogenous levels of human EPHA3/EPHA4/EPHA5 only when phosphorylated at tyrosine 779/833.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.