Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPRS Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89130028 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-028 Supplier Abnova Corporation Supplier No. PAB28518
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant EPRS.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species. Alternative splicing has been observed for this gene, but the full-length nature and biological validity of the variant have not been determined. [provided by RefSeq

Specifications

Antigen EPRS
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant EPRS.
Dilution Western Blot (1:20000) Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene EPRS
Gene Alias DKFZp313B047/EARS/GLUPRORS/PARS/PIG32/QARS/QPRS
Gene Symbols EPRS
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 573-889 of human EPRS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2058
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.