Learn More
ERBB2IP Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERBB2IP.
Supplier: Abnova Corporation PAB5349
Description
This gene is a member of the leucine-rich repeat and PDZ domain (LAP) family. The encoded protein contains 17 leucine-rich repeats and one PDZ domain. It binds to the unphosphorylated form of the ERBB2 protein and regulates ERBB2 function and localization. It has also been shown to affect the Ras signaling pathway by disrupting Ras-Raf interaction. Alternate transcriptional splice variants encoding different isoforms have been found for this gene, but only two of them have been characterized to date. [provided by RefSeq
Specifications
| ERBB2IP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ERBB2IP. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| ERBIN/LAP2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid | 
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| ERBB2IP | |
| ERBB2IP | |
| A synthetic peptide corresponding to N-terminus of human ERBB2IP. | |
| RUO | |
| 55914 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only
