Learn More
ERBB4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERBB4.
Supplier: Abnova Corporation PAB16287
Description
This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily. It encodes a single-pass type I membrane protein with multiple cysteine rich domains, a transmembrane domain, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. Mutations in this gene have been associated with cancer. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq
Specifications
ERBB4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ERBB4. | |
In PBS (0.09% sodium azide) | |
Q15303 | |
ERBB4 | |
A synthetic peptide (conjugated with KLH) corresponding to human ERBB4. | |
50 μg | |
Primary | |
Internal domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-30 ug/mL) The optimal working dilution should be determined by the end user. | |
ERBB4 | |
HER4/MGC138404/p180erbB4 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2066 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only