Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erc1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Erc1.
Supplier: Abnova Corporation PAB15779
Specifications
Erc1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Erc1. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
AK122437 | |
Erc1 | |
Recombinant GST fusion protein corresponding to 192 amino acids of mouse Erc1. | |
RUO | |
111173 | |
Human, Mouse | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Erc1 | |
4930404L01Rik/5033405M01Rik/9630025C19Rik/B430107L16Rik/ELKS/RAB6IP2A/RAB6IP2B/Rab6ip2/mKIAA1081 | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX2134. This antibody detects mERC1 protein. It also recognizes human ERC1 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction