Learn More
ERCC4 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERCC4.
Supplier: Abnova Corporation PAB26907
Description
The protein encoded by this gene forms a complex with ERCC1 and is involved in the 5' incision made during nucleotide excision repair. This complex is a structure specific DNA repair endonuclease that interacts with EME1. Defects in this gene are a cause of xeroderma pigmentosum complementation group F (XP-F), or xeroderma pigmentosum VI (XP6)
Specifications
ERCC4 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
ERCC4 | |
RAD1/XPF | |
Rabbit | |
Affinity purification | |
RUO | |
2072 | |
Human, Mouse | |
Liquid |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ERCC4. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Q92889 | |
ERCC4 | |
A synthetic peptide corresponding to human ERCC4. | |
100 μL | |
Primary | |
ERCC4 polyclonal antibody detects endogenous levels of ERCC4 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only